Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
Plastic bottle
Catalog number FSA217110010
Price (MYR)
440.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
440.00
Add to cart
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Elemental AnalysisPalladium Pd: 9.0-11.0% (dry weight basis)
FormPowder
Loss on Drying45-65%
Biological buffer
RUO – Research Use Only