Acetic acid, potassium salt, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol
Have Questions?
Change view
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA217105000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetic acid, potassium salt
Name NoteFor analysis ACS
pH7.5 to 8.5 (5% soln. at 20°C)
CAS127-08-2
Melting Point292.0°C