Boric acid, 99.5%, for analysis, Thermo Scientific Chemicals
Boric acid, 99.5%, for analysis, Thermo Scientific Chemicals
Boric acid, 99.5%, for analysis, Thermo Scientific Chemicals
Boric acid, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA217085000
Price (MYR)
354.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
354.00
Add to cart
Chemical Identifiers
CAS108-30-5
IUPAC Nameoxolane-2,5-dione
Molecular FormulaC4H4O3
InChI KeyRINCXYDBBGOEEQ-UHFFFAOYSA-N
SMILESO=C1CCC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or flakes and/or chips
Appearance (Color)White
Infrared spectrumConforms
Melting point116°C to 120°C
Titration by coupling>=98.5 %
Biological buffer, Lewis acid
RUO – Research Use Only