Brij™ 30, Thermo Scientific Chemicals
Brij™ 30, Thermo Scientific Chemicals
Brij™ 30, Thermo Scientific Chemicals
Brij™ 30, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Brij™ 30, Thermo Scientific Chemicals
CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number FSA216725000
Price (MYR)
541.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
541.00
Add to cart
Chemical Identifiers
CAS25152-52-7
IUPAC Namealumanylidynestibane
Molecular FormulaAlSb
InChI KeyLVQULNGDVIKLPK-UHFFFAOYSA-N
SMILES[Al]#[Sb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.