N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97%, Thermo Scientific Chemicals
N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97%, Thermo Scientific Chemicals
N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97%, Thermo Scientific Chemicals
N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97%, Thermo Scientific Chemicals
CAS: 1760-24-3 | C8H22N2O3Si | 222.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA216531000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS40876-98-0
IUPAC Namesodium 1,4-diethoxy-1,3,4-trioxobutan-2-ide
Molecular FormulaC8H11NaO5
InChI KeyJPTKZRPOIUYFTM-UHFFFAOYSA-N
SMILES[Na+].CCOC(=O)[CH-]C(=O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow or pale orange to brown or cream
Assay (Non-aqueous acid-base Titration)≥94.0%
FormCrystals or powder or crystalline powder