Chlorotriethylsilane, 99%, Thermo Scientific Chemicals
Chlorotriethylsilane, 99%, Thermo Scientific Chemicals
Chlorotriethylsilane, 99%, Thermo Scientific Chemicals
Chlorotriethylsilane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorotriethylsilane, 99%, Thermo Scientific Chemicals
CAS: 994-30-9 | C6H15ClSi | 150.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA215940250
Price (MYR)
699.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
699.00
Add to cart
Chemical Identifiers
CAS28188-41-2
IUPAC Name3-(bromomethyl)benzonitrile
Molecular FormulaC8H6BrN
InChI KeyCVKOOKPNCVYHNY-UHFFFAOYSA-N
SMILESBrCC1=CC=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)90.5-96.5?C
Silylation reagent
RUO – Research Use Only