Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific Chemicals
Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific Chemicals
Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific Chemicals
Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopentadienyltitanium trichloride, 99%, Thermo Scientific Chemicals
CAS: 1270-98-0 | C5H5Cl3Ti | 219.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number FSA215770025
Price (MYR)
696.00
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
696.00
Add to cart
Chemical Identifiers
CAS1066-51-9
IUPAC Name(aminomethyl)phosphonic acid
Molecular FormulaCH6NO3P
InChI KeyMGRVRXRGTBOSHW-UHFFFAOYSA-N
SMILESNCP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)White