Triton™ X-100, Thermo Scientific™
Triton™ X-100, Thermo Scientific™
Triton™ X-100, Thermo Scientific™
Triton™ X-100, Thermo Scientific™
Triton™ X-100, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number FSA215682500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTriton™ X-100
CAS9002-93-1
Health Hazard 1Danger
Boiling Point270.0°C
Health Hazard 2GHS H Statement
Harmful to aquatic life with long lasting effects.