Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number FSA215370250
Price (MYR)
462.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
462.00
Add to cart
Chemical Identifiers
CAS24974-73-0
IUPAC Name(3-chlorophenyl)methanesulfonyl chloride
Molecular FormulaC7H6Cl2O2S
InChI KeyLXTGNVLBPVVMSL-UHFFFAOYSA-N
SMILESClC1=CC=CC(CS(Cl)(=O)=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Melting Point (clear melt)68.5-77.5?C
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥95.0 to ≤105.0%