Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid, monosodium salt, 99%, pure, anhydrous, Thermo Scientific Chemicals
CAS: 18996-35-5 | C6H7NaO7 | 214.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA215331000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS8002-03-7
SpecificationsSpecification SheetSpecification Sheet
Unsaponifiable matter=<1 %
Composition of fatty acids=<0.4 % (GC) Saturated of chain length
Composition of fatty acids5.0 to 14.0 % (GC) Palmitic acid
Composition of fatty acids1.3 to 6.5 % (GC) Stearic acid
Composition of fatty acids35.0 to 76.0 % (GC) Oleic acid