Potassium indigotrisulfonate
Potassium indigotrisulfonate
Potassium indigotrisulfonate
Potassium indigotrisulfonate
Thermo Scientific Chemicals
Potassium indigotrisulfonate
CAS: 67627-18-3 | C16H7K3N2O11S3 | 616.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA215170010
Price (MYR)
644.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
644.00
Add to cart
Chemical Identifiers
CAS6915-15-7
IUPAC Name2-hydroxybutanedioic acid
Molecular FormulaC4H6O5
InChI KeyBJEPYKJPYRNKOW-UHFFFAOYNA-N
SMILESOC(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5%
Optical Rotation-0.5? to +0.5? (c= 8.5 in water)
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%