Chromoxane Cyanine R, pure
Chromoxane Cyanine R, pure
Chromoxane Cyanine R, pure
Chromoxane Cyanine R, pure
Thermo Scientific Chemicals
Chromoxane Cyanine R, pure
CAS: 3564-18-9 | C23H15Na3O9S
Have Questions?
Catalog NumberQuantity
FSA2151500505 g
Catalog number FSA215150050
Price (MYR)
254.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
254.00
Add to cart
Specifications
Chemical Name or MaterialChromoxane Cyanine R
CAS3564-18-9
Molecular FormulaC23H15Na3O9S
MDL NumberMFCD00012411
Beilstein11, II, 240
View more
Indicator
RUO – Research Use Only