Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals
Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals
Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals
Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorhexidine diacetate hydrate, 98%, Thermo Scientific Chemicals
CAS: 206986-79-0 | C26H38Cl2N10O4 | 625.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA214980250
Price (MYR)
931.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
931.00
Add to cart
Chemical Identifiers
CAS431-03-8
IUPAC Namebutane-2,3-dione
Molecular FormulaC4H6O2
InChI KeyQSJXEFYPDANLFS-UHFFFAOYSA-N
SMILESCC(=O)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.3930-1.3980 @ 20?C
Assay (GC)≥98.5%