Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals
Molecular sieves 4A, powder &lt;50 micron, Thermo Scientific Chemicals
Molecular sieves 4A, powder &lt;50 micron, Thermo Scientific Chemicals
Molecular sieves 4A, powder &lt;50 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic Bottle
Catalog number FSA214805000
Price (MYR)
553.00
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
553.00
Add to cart
Specifications
Chemical Name or MaterialMolecular sieves pack 4A beads,8-12 mesh
ColorWhite to Beige
PackagingPlastic Bottle
Residual Water3% max.
Water Adsorption18% max.
View more
Drying agent
RUO – Research Use Only