Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals
Molecular sieves 4A, powder &lt;50 micron, Thermo Scientific Chemicals
Molecular sieves 4A, powder &lt;50 micron, Thermo Scientific Chemicals
Molecular sieves 4A, powder &lt;50 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
Plastic Bottle
Catalog number FSA214800010
Price (MYR)
844.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
844.00
Add to cart
Chemical Identifiers
CAS144896-51-5
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤2.5%
Refractive Index1.5825-1.5885 @ 20?C
Appearance (Color)Clear yellow
Assay (GC)≥94.0%
FormLiquid
Drying agent
RUO – Research Use Only