3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
CAS: 580-17-6 | C9H8N2 | 144.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA213780010
Price (MYR)
310.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
310.00
Add to cart
Specifications
Chemical Name or Material3-Aminoquinoline
Name Note99+%
CAS580-17-6
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning