Iodine, 99.5+%, for analysis
Iodine, 99.5+%, for analysis
Iodine, 99.5+%, for analysis
Iodine, 99.5+%, for analysis
Thermo Scientific Chemicals
Iodine, 99.5+%, for analysis
CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA21249025025 gGlass bottle
Catalog number FSA212490250
Price (MYR)
259.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
259.00
Add to cart
Chemical Identifiers
CAS70-11-1
IUPAC Name2-bromo-1-phenylethan-1-one
Molecular FormulaC8H7BrO
InChI KeyLIGACIXOYTUXAW-UHFFFAOYSA-N
SMILESBrCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point46°C to 51°C
GC>=97.5 %
Appearance (Color)White to green-brown