Iron(III) citrate hydrate, 98%
Iron(III) citrate hydrate, 98%
Iron(III) citrate hydrate, 98%
Iron(III) citrate hydrate, 98%
Thermo Scientific Chemicals
Iron(III) citrate hydrate, 98%
CAS: 334024-15-6 | C6H5FeO7 | 244.94 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA2118500101 kgPlastic bottle
Catalog number FSA211850010
Price (MYR)
880.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
880.00
Add to cart
Chemical Identifiers
CAS71-41-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
GC>=98.5 %
Water=<0.2 %