Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
CAS: 94-36-0 | C14H10O4 | 242.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA211780050
Price (MYR)
281.00
Add to cart
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
281.00
Add to cart
Chemical Identifiers
CAS3032-92-6
IUPAC Name4-ethynylbenzonitrile
Molecular FormulaC9H5N
InChI KeyLAGNMUUUMQJXBF-UHFFFAOYSA-N
SMILESC#CC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥96.0% (GC)
Appearance (Color)White to dark brown
FormPowder or crystals
Radical initiators
RUO – Research Use Only