Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibenzoyl peroxide, 75%, remainder water, Thermo Scientific Chemicals
CAS: 94-36-0 | C14H10O4 | 242.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
100 g
Packaging:
Plastic bottle
Catalog number FSA211780010
Price (MYR)
1,111.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,111.00
Add to cart
Specifications
Chemical Name or MaterialDibenzoyl peroxide
Name Note75%, remainder water
CAS94-36-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Heating may cause a fire.
Very toxic to aquatic life with long lasting effects.
View more
Radical initiators
RUO – Research Use Only