Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals
CAS: 92112-69-1 | C6H14
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
Packaging:
Metal Drum
Catalog number FSA210590250
Price (MYR)
1,515.00
Add to cart
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Price (MYR)
1,515.00
Add to cart
Chemical Identifiers
CAS93983-13-2
IUPAC Name(2-chloro-3,4-dimethoxyphenyl)methanol
Molecular FormulaC9H11ClO3
InChI KeyZXYBQLFOEYWYBM-UHFFFAOYSA-N
SMILESCOC1=CC=C(CO)C(Cl)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay (GC)≥96.0%
Melting Point (clear melt)67.5-73.5?C