N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
500 mL
1 L
2.5 L
25 L
Packaging:
View all
Plastic Drum
Catalog number FSA210580100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS13446-34-9
IUPAC Namemanganese(2+) tetrahydrate dichloride
Molecular FormulaCl2H8MnO4
InChI KeyCNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILESO.O.O.O.[Cl-].[Cl-].[Mn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<50 ppm
Iron (Fe)=<5 ppm
Magnesium (Mg)=<50 ppm
KMnO4-reducing substance=<0.0005 % (as O)
Titration Complexometric>=99.0 %
View more
For analysis
RUO – Research Use Only