Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
CAS: 12501-23-4 | H5O41PW12 | 2880.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA208311000
Price (MYR)
1,154.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,154.00
Add to cart
Chemical Identifiers
CAS62961-64-2
IUPAC Name1,4-bis(propan-2-yl) (2R,3S)-2,3-dihydroxybutanedioate
Molecular FormulaC10H18O6
InChI KeyXEBCWEDRGPSHQH-OCAPTIKFSA-N
SMILESCC(C)OC(=O)[C@@H](O)[C@@H](O)C(=O)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4375-1.4415 @ 20?C
Appearance (Color)Clear colorless
Optical Rotation-15.5 ± 1.0? (neat)
FormLiquid or viscous liquid