CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
Thermo Scientific Chemicals
CHES, 98+%, for biochemistry
CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA208185000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Absorbance(0.1 M aq. soln.)
Chemical Name or MaterialCHES
Absorbance Conditions(0.1M aq. soln.)
Name Note98+%
CAS103-47-9
View more
Biological buffer
RUO – Research Use Only