CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
Thermo Scientific Chemicals
CHES, 98+%, for biochemistry
CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA208180250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS32387-21-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder
Melting point197°C to 202°C
Appearance (Color)White to beige
Infrared spectrumConforms
HPLC>=96.0 %
View more
Biological buffer
RUO – Research Use Only