Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Thermo Scientific Chemicals
Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ascarite™ II, CO2 absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Thermo Scientific Chemicals
CAS: 81133-20-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Catalog number FSA208080025
Price (MYR)
12,645.00
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (MYR)
12,645.00
Add to cart
Chemical Identifiers
CAS66-77-3
IUPAC Namenaphthalene-1-carbaldehyde
Molecular FormulaC11H8O
InChI KeySQAINHDHICKHLX-UHFFFAOYSA-N
SMILESO=CC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.6500-1.6550 @ 20?C
Appearance (Color)Clear pale yellow to yellow-brown
CommentTrace acid content may precipitate at low temperatures - re-dissolves on warming
Gas Adsorbent
RUO – Research Use Only