Copper, 99.9%, Thermo Scientific Chemicals
Copper, 99.9%, Thermo Scientific Chemicals
Copper, 99.9%, Thermo Scientific Chemicals
Copper, 99.9%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper, 99.9%, Thermo Scientific Chemicals
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Catalog number FSA207695000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight63.54
GradeTrace Metal Basis
Unit Size
For ultra trace analysis
RUO – Research Use Only