Spironolactone, 99%
Spironolactone, 99%
Spironolactone, 99%
Thermo Scientific Chemicals
Spironolactone, 99%
CAS: 52-01-7 | C24H32O4S | 416.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA207460050
Price (MYR)
1,436.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,436.00
Add to cart
Chemical Identifiers
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentBoron, B: ≤0.1ppm (≤100ppb)
Water Content (Karl Fischer Titration)≤0.02%
Trace MetalAluminum, Al: ≤0.1ppm (≤100ppb)
Trace MetalArsenic, As, and Antimony, Sb (as Arsenic); ≤0.01ppm (≤10ppb)
Trace MetalCalcium, Ca: ≤0.1ppm (≤100ppb)