1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals
1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals
1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals
1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Phenyl-2-thiourea, 97%, Thermo Scientific Chemicals
CAS: 103-85-5 | C7H8N2S | 152.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA207250050
Price (MYR)
281.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
281.00
Add to cart
Chemical Identifiers
CAS50606-95-6
IUPAC Name4-hexylbenzoyl chloride
Molecular FormulaC13H17ClO
InChI KeyXRAHLPNMIIAEPP-UHFFFAOYSA-N
SMILESCCCCCCC1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5235-1.5275 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%