L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals
L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals
L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals
L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(-)-Phenylephrine hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 61-76-7 | C9H14ClNO2 | 203.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Catalog number FSA207240100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS5470-11-1
IUPAC Namehydroxylamine hydrochloride
Molecular FormulaClH4NO
InChI KeyWTDHULULXKLSOZ-UHFFFAOYSA-N
SMILESCl.NO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Melting point151°C to 157°C (Dec)
Titration with NaOH>=96.0 %