Lithium tetraborate, 98+%, for analysis, Thermo Scientific Chemicals
Lithium tetraborate, 98+%, for analysis, Thermo Scientific Chemicals
Lithium tetraborate, 98+%, for analysis, Thermo Scientific Chemicals
Lithium tetraborate, 98+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lithium tetraborate, 98+%, for analysis, Thermo Scientific Chemicals
CAS: 12007-60-2 | B4Li2O7 | 169.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA206942500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)≥99%
Water Content≤0.05%
CommentSpecification differs for U.S. and non-U.S. material where indicated
CommentSubmicron filtered
Optical Absorbance at215 nm: ≤1.00 AU [U.S. specification]