Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 7782-68-5 | ILiO3 | 181.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA206450500
Price (MYR)
562.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
562.00
Add to cart
Chemical Identifiers
CAS98327-87-8
IUPAC Name[2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane
Molecular FormulaC44H32P2
InChI KeyMUALRAIOVNYAIW-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Melting point280°C to 285°C
Oxidation reagent for analysis
RUO – Research Use Only