Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number FSA206381000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS766-39-2
IUPAC Namedimethyl-2,5-dihydrofuran-2,5-dione
Molecular FormulaC6H6O3
InChI KeyMFGALGYVFGDXIX-UHFFFAOYSA-N
SMILESCC1=C(C)C(=O)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Titration with TBAH>=96.0 %
Appearance (Color)White to beige
Melting point91°C to 96°C
Oxidation reagent
RUO – Research Use Only