Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphomolybdic acid hydrate, 80%, Thermo Scientific Chemicals
CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA206380250
Price (MYR)
549.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
549.00
Add to cart
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Fibers
Oxidation reagent
RUO – Research Use Only