Ammonium nitrate, 99+%, for analysis
Product Image
Thermo Scientific Chemicals
Ammonium nitrate, 99+%, for analysis
Have Questions?
Catalog number FSA205860025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS109-02-4
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.4340-1.4370 @ 20?C
  • Ammonium nitrate, 99+%, for analysis 2.5kg Plastic Bottle Cas No. 6484-52-2
RUO – Research Use Only