2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals
2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals
2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals
2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Thiophenemethylamine, 95%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Catalog number FSA205570250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS6781-42-6
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Melting Point (clear melt)29.0-38.0?C
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or fused solid