Leuco Crystal Violet, 99%, pure
Leuco Crystal Violet, 99%, pure
Leuco Crystal Violet, 99%, pure
Leuco Crystal Violet, 99%, pure
Thermo Scientific Chemicals
Leuco Crystal Violet, 99%, pure
CAS: 603-48-5 | C25H31N3 | 373.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA204330250
Price (MYR)
567.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
567.00
Add to cart
Chemical Identifiers
CAS3061-36-7
IUPAC Name1,4-diphenoxybenzene
Molecular FormulaC18H14O2
InChI KeyUVGPELGZPWDPFP-UHFFFAOYSA-N
SMILESO(C1=CC=CC=C1)C1=CC=C(OC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale pink
Melting Point (clear melt)68-77?C
FormPowder or flakes/pellets/prills or lumps
Assay (GC)≥96.0%