Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, >-97.5% (on dry substance), C3HCl2N3NaO3, CAS Number-2893-78-9, sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt, 500g, 98%, White, 220-767-7, 219.95, Danger
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Catalog number FSA204115000
Price (MYR)
470.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
470.00
Add to cart
Chemical Identifiers
CAS33018-91-6
IUPAC Name7-ethoxy-7-oxoheptanoic acid
Molecular FormulaC9H16O4
InChI KeyNQYXFXWKKYGBNL-UHFFFAOYSA-N
SMILESCCOC(=O)CCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Refractive Index1.4400-1.4440 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
C-X bond formation reagent, Oxidation reagent
RUO – Research Use Only