Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 98%, Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, >-97.5% (on dry substance), C3HCl2N3NaO3, CAS Number-2893-78-9, sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt, 500g, 98%, White, 220-767-7, 219.95, Danger
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Catalog number FSA204110250
Price (MYR)
249.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
249.00
Add to cart
Specifications
Chemical Name or MaterialDichloroisocyanuric acid sodium salt
Name Note98%
CAS2893-78-9
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.
May intensify fire; oxidizer.
Contact with acids liberates toxic gas.
View more
C-X bond formation reagent, Oxidation reagent
RUO – Research Use Only