Cyclophosphamide monohydrate, 97%, Thermo Scientific Chemicals
Cyclophosphamide monohydrate, 97%, Thermo Scientific Chemicals
Cyclophosphamide monohydrate, 97%, Thermo Scientific Chemicals
Cyclophosphamide monohydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclophosphamide monohydrate, 97%, Thermo Scientific Chemicals
CAS: 6055-19-2 | C7H17Cl2N2O3P | 279.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA203960010
Price (MYR)
395.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
395.00
Add to cart
Chemical Identifiers
CAS546-89-4
IUPAC Namelithium(1+) acetate
Molecular FormulaC2H3LiO2
InChI KeyXIXADJRWDQXREU-UHFFFAOYSA-M
SMILES[Li+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=99.0 %
Melting point280°C to 285°C