alpha-Terpineol, 95%, mixture of alpha- and gamma-terpineol, Thermo Scientific Chemicals
alpha-Terpineol, 95%, mixture of alpha- and gamma-terpineol, Thermo Scientific Chemicals
alpha-Terpineol, 95%, mixture of alpha- and gamma-terpineol, Thermo Scientific Chemicals
alpha-Terpineol, 95%, mixture of alpha- and gamma-terpineol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
alpha-Terpineol, 95%, mixture of alpha- and gamma-terpineol, Thermo Scientific Chemicals
CAS: 98-55-5 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA203740500
Price (MYR)
287.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
287.00
Add to cart
Chemical Identifiers
CAS16712-20-2
IUPAC Namelithium(1+) hydrate chloride
Molecular FormulaClH2LiO
InChI KeyVXJIMUZIBHBWBV-UHFFFAOYSA-M
SMILES[Li+].O.[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.