(-)-Limonene, 96%, Thermo Scientific Chemicals
(-)-Limonene, 96%, Thermo Scientific Chemicals
(-)-Limonene, 96%, Thermo Scientific Chemicals
(-)-Limonene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(-)-Limonene, 96%, Thermo Scientific Chemicals
CAS: 5989-54-8 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA203735000
Price (MYR)
916.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
916.00
Add to cart
Chemical Identifiers
CAS100-19-6
IUPAC Name1-(4-nitrophenyl)ethan-1-one
Molecular FormulaC8H7NO3
InChI KeyYQYGPGKTNQNXMH-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Free acid=<0.2 % (as 4-Nitrobenzoic acid)
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Crystalline powder