Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical, Thermo Scientific Chemicals
CAS: 584-84-9 | C9H6N2O2 | 174.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA202500025
Price (MYR)
2,359.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,359.00
Add to cart
Chemical Identifiers
CAS330804-03-0
IUPAC Name(2-methanesulfonylphenyl)boronic acid
Molecular FormulaC7H9BO4S
InChI KeyMSQFFCRGQPVQRS-UHFFFAOYSA-N
SMILESCS(=O)(=O)C1=CC=CC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%