4-(2-Aminoethyl)pyridine, 98%
4-(2-Aminoethyl)pyridine, 98%
4-(2-Aminoethyl)pyridine, 98%
4-(2-Aminoethyl)pyridine, 98%
Thermo Scientific Chemicals
4-(2-Aminoethyl)pyridine, 98%
CAS: 13258-63-4 | C7H10N2 | 122.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA2020300101 gGlass bottle
Catalog number FSA202030010
Price (MYR)
1,070.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,070.00
Add to cart
Specifications
Chemical Name or Material4-(2-Aminoethyl)pyridine
CAS13258-63-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.