Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
FSA201672500250 gPlastic bottle
Catalog number FSA201672500
Price (MYR)
774.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
774.00
Add to cart
Chemical Identifiers
CAS5351-23-5
IUPAC Name4-hydroxybenzohydrazide
Molecular FormulaC7H8N2O2
InChI KeyZMZGIVVRBMFZSG-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH>=97.5 %
Appearance (Color)White to light beige
Infrared spectrumConforms
For ultra trace analysis
RUO – Research Use Only