Disperse Orange 3, 65%
Disperse Orange 3, 65%
Disperse Orange 3, 65%
Disperse Orange 3, 65%
Thermo Scientific Chemicals
Disperse Orange 3, 65%
CAS: 730-40-5 | C12H10N4O2 | 242.238 g/mol
Have Questions?
Catalog NumberQuantity
FSA20162050050 g
Catalog number FSA201620500
Price (MYR)
2,021.00
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
2,021.00
Add to cart
Chemical Identifiers
CAS7440-58-6
IUPAC Namehafnium
Molecular FormulaHf
InChI KeyVBJZVLUMGGDVMO-UHFFFAOYSA-N
SMILES[Hf]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace MetalTotal metal impurities less than 0.05% excluding Zr, Zr nominal 3%