Pararosaniline base, Thermo Scientific Chemicals
Pararosaniline base, Thermo Scientific Chemicals
Pararosaniline base, Thermo Scientific Chemicals
Pararosaniline base, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pararosaniline base, Thermo Scientific Chemicals
CAS: 467-62-9 | C19H19N3O | 305.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Catalog number FSA201550100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS55499-43-9
IUPAC Name(3,4-dimethylphenyl)boronic acid
Molecular FormulaC8H11BO2
InChI KeyKDVZJKOYSOFXRV-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1C)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥98.0%
Proton NMRConforms to structure