Zinc, +99%, mossy, Thermo Scientific Chemicals
Zinc, +99%, mossy, Thermo Scientific Chemicals
Zinc, +99%, mossy, Thermo Scientific Chemicals
Zinc, +99%, mossy, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Zinc, +99%, mossy, Thermo Scientific Chemicals
CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA201455000
Price (MYR)
371.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
371.00
Add to cart
Chemical Identifiers
CAS112-89-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Melting Point (clear melt)25.0-34.0?C
FormFused solid
Assay (GC)≥96.0%
Reduction reagent
RUO – Research Use Only