1-Methyladenine, 98+%
1-Methyladenine, 98+%
1-Methyladenine, 98+%
1-Methyladenine, 98+%
Thermo Scientific Chemicals
1-Methyladenine, 98+%
CAS: 5142-22-3 | C6H7N5 | 149.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
250 mg
Packaging:
Glass bottle
Catalog number FSA201310250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Methyladenine
CAS5142-22-3
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0