Sulfur, 99.5+%, refined, Thermo Scientific Chemicals
Sulfur, 99.5+%, refined, Thermo Scientific Chemicals
Sulfur, 99.5+%, refined, Thermo Scientific Chemicals
Sulfur, 99.5+%, refined, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfur, 99.5+%, refined, Thermo Scientific Chemicals
CAS: 7704-34-9 | S | 32.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA201225000
Price (MYR)
398.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
398.00
Add to cart
Chemical Identifiers
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.005 % (Coulometric)
Appearance (Color)Clear colorless
GC>=99.8 %
Acidity (HCl)=<0.002 %
View more
For ultra trace analysis
RUO – Research Use Only